The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_005308.2 | NP_005299.1 | 3 - 4 | 496 | 85 |
XM_005269707.1 | XP_005269764.1 | 3 - 4 | 493 | 85 | |
XM_005269708.1 | XP_005269765.1 | 2 - 3 | 386 | 85 | |
GenBank mRNA | AK298261.1 | - | 3 - 4 | 439 | 85 |
AK301450.1 | - | 2 - 3 | 187 | 85 | |
AK313012.1 | - | 3 - 4 | 343 | 85 | |
BC064506.1 | - | 3 - 4 | 490 | 85 | |
L15388.1 | - | 3 - 4 | 483 | 85 |