The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_007191.4 | NP_009122.2 | 6 - 7 | 1108 | 72 |
GenBank mRNA | AF122922.1 | - | 6 - 7 | 845 | 72 |
AK301818.1 | - | 6 - 7 | 1035 | 72 | |
AY358344.1 | - | 6 - 7 | 878 | 72 | |
BC018037.1 | - | 6 - 7 | 851 | 72 | |
BX647427.1 | - | 6 - 7 | 777 | 72 |