The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_001077196.1 | NP_001070664.1 | 15 - 16 | 1385 | 60 |
NM_001077197.1 | NP_001070665.1 | 19 - 20 | 1915 | 60 | |
NM_001077358.1 | NP_001070826.1 | 17 - 18 | 1625 | 60 | |
NM_016953.3 | NP_058649.3 | 18 - 19 | 2884 | 60 | |
GenBank mRNA | AB036704.1 | - | 18 - 19 | 2884 | 60 |
AB038041.1 | - | 19 - 20 | 1915 | 60 | |
AF281865.1 | - | 17 - 18 | 1597 | 60 | |
AJ251509.1 | - | 15 - 16 | 1384 | 60 | |
AJ278682.1 | - | 19 - 20 | 1958 | 60 | |
AK074273.1 | - | 8 - 9 | 782 | 60 | |
BC112393.1 | - | 18 - 19 | 2875 | 60 | |
BC114431.1 | - | 18 - 19 | 2875 | 60 | |
HM771389.1 | - | 18 - 19 | 2566 | 60 | |
HM771390.1 | - | 18 - 19 | 2566 | 60 | |
HM771391.1 | - | 18 - 19 | 2566 | 60 |