Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
AOC3
Entrez Gene ID
8639
Gene Name
amine oxidase, copper containing 3
Gene Aliases
HPAO, SSAO, VAP-1, VAP1
Chromosome Location
Chr.17: 42851123 - 42858130 on Build GRCh38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001277732.1 NP_001264661.1 2 - 3 413 58
NM_003734.3 NP_003725.1 2 - 3 2051 58
NR_102422.1 2 - 3 1962 58
XM_011525419.1 XP_011523721.1 3 - 4 2205 58
GenBank mRNA AF067406.1 - 2 - 3 1971 58
AK025727.1 - 2 - 3 541 58
AK315129.1 - 2 - 3 2030 58
BC050549.1 - 2 - 3 2001 58
DA592022.1 - 2 - 3 413 58
DB250074.1 - 2 - 3 393 58
U39447.1 - 2 - 3 2051 58

View Full Product Details