The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_001081955.2 | NP_001075424.1 | 12 - 13 | 1052 | 72 |
NM_001165933.1 | NP_001159405.1 | 12 - 13 | 1052 | 72 | |
NM_003835.3 | NP_003826.2 | 12 - 13 | 1061 | 72 | |
XM_011525426.2 | XP_011523728.1 | 5 - 6 | 1232 | 72 | |
GenBank mRNA | AF071476.1 | - | 12 - 13 | 925 | 72 |
AF073710.1 | - | 3 - 4 | 786 | 72 | |
AF493932.1 | - | 2 - 3 | 171 | 72 | |
AF493933.1 | - | 12 - 13 | 858 | 72 | |
AK289876.1 | - | 12 - 13 | 925 | 72 | |
AK290535.1 | - | 12 - 13 | 921 | 72 | |
AK301719.1 | - | 12 - 13 | 931 | 72 | |
AK309061.1 | - | 12 - 13 | 942 | 72 | |
AY585190.1 | - | 2 - 3 | 171 | 72 | |
AY585191.1 | - | 12 - 13 | 849 | 72 | |
BC096843.1 | - | 2 - 3 | 174 | 72 |