The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_004186.4 | NP_004177.3 | 6 - 7 | 751 | 108 |
XM_005265381.4 | XP_005265438.1 | 6 - 7 | 881 | 108 | |
XM_006713290.3 | XP_006713353.1 | 6 - 7 | 684 | 108 | |
XM_011533998.2 | XP_011532300.1 | 6 - 7 | 683 | 108 | |
XM_011534000.2 | XP_011532302.1 | 6 - 7 | 684 | 108 | |
GenBank mRNA | BC042914.1 | - | 6 - 7 | 747 | 108 |
U33920.1 | - | 5 - 6 | 627 | 108 |