The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_005895.3 | NP_005886.2 | 18 - 19 | 4144 | 83 |
XM_005266164.4 | XP_005266221.1 | 18 - 19 | 3751 | 83 | |
XM_005266167.4 | XP_005266224.1 | 15 - 16 | 3176 | 83 | |
XM_006719736.3 | XP_006719799.1 | 18 - 19 | 3871 | 83 | |
XM_006719737.3 | XP_006719800.1 | 18 - 19 | 3881 | 83 | |
XM_011534793.2 | XP_011533095.1 | 17 - 18 | 3598 | 83 | |
XM_011534794.2 | XP_011533096.1 | 10 - 11 | 1749 | 83 | |
GenBank mRNA | AB027133.1 | - | 18 - 19 | 3866 | 83 |
AF485338.1 | - | 17 - 18 | 3588 | 83 | |
D63997.1 | - | 18 - 19 | 3854 | 83 | |
L06148.1 | - | 8 - 9 | 1278 | 83 |