The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_001256618.1 | NP_001243547.1 | 4 - 5 | 799 | 96 |
NM_006659.3 | NP_006650.1 | 5 - 6 | 988 | 96 | |
NR_046330.1 | 5 - 6 | 1372 | 96 | ||
GenBank mRNA | AF042379.1 | - | 5 - 6 | 678 | 96 |
AK092113.1 | - | 5 - 6 | 1372 | 96 | |
AK223586.1 | - | 5 - 6 | 879 | 96 | |
AK297251.1 | - | 4 - 5 | 521 | 96 | |
AK299564.1 | - | 5 - 6 | 542 | 96 | |
AK314520.1 | - | 5 - 6 | 710 | 96 | |
BC093770.1 | - | 4 - 5 | 667 | 96 | |
BC111957.1 | - | 4 - 5 | 667 | 96 |