The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_014323.2 | NP_055138.2 | 3 - 4 | 2168 | 77 |
GenBank mRNA | AF254085.1 | - | 3 - 4 | 2137 | 77 |