Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
NUDCD3
Entrez Gene ID
23386
Gene Name
NudC domain containing 3
Gene Aliases
NudCL
Chromosome Location
Chr.7: 44382366 - 44490875 on Build GRCh38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_015332.3 NP_056147.2 3 - 4 832 63
XM_011515247.2 XP_011513549.1 3 - 4 921 63
XM_017011909.1 XP_016867398.1 4 - 5 858 63
XM_017011910.1 XP_016867399.1 4 - 5 716 63
GenBank mRNA AB028991.1 - 3 - 4 626 63
AK024226.1 - 2 - 3 340 63
BC003691.2 - 3 - 4 693 63
BC011673.2 - 2 - 3 452 63

View Full Product Details