The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_015388.3 | NP_056203.2 | 3 - 4 | 578 | 120 |
XM_005248989.4 | XP_005249046.1 | 3 - 4 | 614 | 120 | |
XM_005248990.2 | XP_005249047.1 | 3 - 4 | 619 | 120 | |
XM_011514458.2 | XP_011512760.1 | 3 - 4 | 721 | 120 | |
GenBank mRNA | AF162672.1 | - | 3 - 4 | 493 | 120 |
AK000946.1 | - | 3 - 4 | 514 | 120 | |
AK021433.1 | - | - | 490 | 120 | |
AK021655.1 | - | 3 - 4 | 458 | 120 | |
AK022757.1 | - | 3 - 4 | 477 | 120 | |
AK298150.1 | - | 3 - 4 | 614 | 120 | |
AL050274.1 | - | 3 - 4 | 379 | 120 | |
BC019297.1 | - | 3 - 4 | 527 | 120 | |
CR533541.1 | - | 2 - 3 | 290 | 120 |