The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_015559.2 | NP_056374.2 | 4 - 5 | 4298 | 80 |
GenBank mRNA | AB007897.2 | - | 4 - 5 | 4298 | 80 |
AB022660.1 | - | 3 - 4 | 3845 | 80 | |
BC146776.1 | - | 4 - 5 | 4298 | 80 | |
BQ641251.1 | - | 1 - 2 | 394 | 80 |