The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_001135955.1 | NP_001129427.1 | 8 - 9 | 2312 | 76 |
NM_001135957.1 | NP_001129429.1 | 8 - 9 | 2312 | 76 | |
NM_001135958.1 | NP_001129430.1 | 7 - 8 | 1793 | 76 | |
NM_016179.2 | NP_057263.1 | 8 - 9 | 2312 | 76 | |
XM_017020724.1 | XP_016876213.1 | 8 - 9 | 2873 | 76 | |
GenBank mRNA | AF063822.1 | - | 8 - 9 | 2312 | 76 |
AF063823.1 | - | 8 - 9 | 2312 | 76 | |
AF063824.1 | - | 8 - 9 | 2312 | 76 | |
AF175406.1 | - | 8 - 9 | 2308 | 76 | |
AF421358.1 | - | 7 - 8 | 2077 | 76 | |
AF421359.1 | - | 7 - 8 | 2077 | 76 | |
AF421361.1 | - | 6 - 7 | 1740 | 76 | |
AF421362.1 | - | 6 - 7 | 1558 | 76 | |
AK312524.1 | - | 8 - 9 | 2282 | 76 | |
BC104725.1 | - | 8 - 9 | 2233 | 76 | |
BC105306.1 | - | 7 - 8 | 1919 | 76 | |
BC105641.1 | - | 8 - 9 | 2233 | 76 | |
BC105917.1 | - | 8 - 9 | 2233 | 76 | |
BX648312.1 | - | 8 - 9 | 2936 | 76 |