The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_017818.3 | NP_060288.3 | 1 - 2 | 179 | 83 |
XM_005244754.1 | XP_005244811.1 | 1 - 2 | 174 | 83 | |
XM_017001387.1 | XP_016856876.1 | 1 - 2 | 174 | 83 | |
GenBank mRNA | AB034912.1 | - | 1 - 2 | 130 | 83 |
AK000437.1 | - | 1 - 2 | 145 | 83 | |
AK225525.1 | - | 1 - 2 | 145 | 83 | |
AK302397.1 | - | 1 - 2 | 175 | 83 | |
BC086311.1 | - | 1 - 2 | 152 | 83 | |
DC380547.1 | - | 1 - 2 | 181 | 83 |