Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
CUEDC1
Entrez Gene ID
404093
Gene Name
CUE domain containing 1
Gene Aliases
-
Chromosome Location
Chr.17: 57861243 - 57955323 on Build GRCh38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001271875.1 NP_001258804.1 3 - 4 880 117
NM_001292025.1 NP_001278954.1 3 - 4 1185 117
XM_006721900.2 XP_006721963.1 3 - 4 1495 117
XM_017024640.1 XP_016880129.1 3 - 4 850 117
GenBank mRNA AK000746.1 - 3 - 4 1185 117
AK000977.1 - 2 - 3 619 117
BC056882.1 - 3 - 4 880 117

View Full Product Details