Assay Design
The assay probe spans an exon junction.
Gene Transcripts
- Gene Symbol
- GSDMB
- Entrez Gene ID
-
55876
- Gene Name
- gasdermin B
- Gene Aliases
-
GSDML, PP4052, PRO2521
- Chromosome Location
-
Chr.17: 39904595 - 39919377 on Build GRCh38
View Full Product Details