Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.22: 26464213-26464704 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
HPS4
Entrez Gene ID
89781
Gene Name
HPS4, biogenesis of lysosomal organelles complex 3 subunit 2
Gene Aliases
BLOC3S2, LE
UniGene
Hs.474436
Gene Chromosome Location
Chr.22: 26443613-26483863 on Build GRCh38
Transcripts
NM_022081.5 NM_152841.2 NR_073135.1 NR_073136.1 XM_006724353.3 XM_006724354.3 XM_006724360.3 XM_011530485.1 XM_011530486.2 XM_011530487.2 XM_011530488.2 XM_011530489.2 XM_011530490.2 XM_011530491.2 XM_011530492.1 XM_011530493.2 XM_011530494.2 XM_011530495.2 XM_011530496.2 XM_017029042.1 XM_017029043.1 XM_017029044.1 XM_017029045.1 XM_017029046.1 XM_017029047.1 XM_017029048.1 XM_017029049.1 XM_017029050.1 XM_017029051.1 XM_017029052.1 XM_017029053.1 XM_017029054.1 XM_017029055.1 XM_017029056.1 XM_017029057.1 XM_017029058.1 XM_017029059.1 XM_017029060.1 XM_017029061.1 XM_017029062.1 XM_017029063.1 XM_017029064.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
1032977 c.985G>A p.E329K Chr.22: 26464684-26464684 on Build GRCh38
4691095 c.1180G>A p.E394K Chr.22: 26464489-26464489 on Build GRCh38
2935545 c.997G>C p.D333H Chr.22: 26464672-26464672 on Build GRCh38
212770 c.1271G>A p.S424N Chr.22: 26464359-26464359 on Build GRCh38
5812621 c.985G>T p.E329* Chr.22: 26464684-26464684 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs759348602 Chr.22: 26464488-26464488 on Build GRCh38
rs143711674 Chr.22: 26464408-26464408 on Build GRCh38
rs773968140 Chr.22: 26464529-26464542 on Build GRCh38
rs868526867 Chr.22: 26464241-26464241 on Build GRCh38
rs996720116 Chr.22: 26464481-26464481 on Build GRCh38
rs758581869 Chr.22: 26464341-26464341 on Build GRCh38
rs117114544 Chr.22: 26464323-26464323 on Build GRCh38
rs200477299 Chr.22: 26464471-26464471 on Build GRCh38
rs372970834 Chr.22: 26464594-26464594 on Build GRCh38
rs750452419 Chr.22: 26464317-26464317 on Build GRCh38
rs186173240 Chr.22: 26464270-26464270 on Build GRCh38
rs764993355 Chr.22: 26464490-26464490 on Build GRCh38
rs142139781 Chr.22: 26464450-26464450 on Build GRCh38
rs772205948 Chr.22: 26464247-26464247 on Build GRCh38
rs776612681 Chr.22: 26464546-26464546 on Build GRCh38
rs202197120 Chr.22: 26464684-26464684 on Build GRCh38
rs749976459 Chr.22: 26464396-26464396 on Build GRCh38
rs983056340 Chr.22: 26464254-26464254 on Build GRCh38
rs2014410 Chr.22: 26464303-26464303 on Build GRCh38
rs376253108 Chr.22: 26464528-26464528 on Build GRCh38
rs760276585 Chr.22: 26464480-26464480 on Build GRCh38
rs771794197 Chr.22: 26464522-26464522 on Build GRCh38
rs369393785 Chr.22: 26464538-26464538 on Build GRCh38
rs759075680 Chr.22: 26464572-26464572 on Build GRCh38
rs772691909 Chr.22: 26464553-26464553 on Build GRCh38
rs386820399 Chr.22: 26464569-26464570 on Build GRCh38
rs371109134 Chr.22: 26464674-26464674 on Build GRCh38
rs765633123 Chr.22: 26464297-26464297 on Build GRCh38
rs964934683 Chr.22: 26464478-26464478 on Build GRCh38
rs775959392 Chr.22: 26464634-26464634 on Build GRCh38
rs753803734 Chr.22: 26464681-26464681 on Build GRCh38
rs746818979 Chr.22: 26464361-26464361 on Build GRCh38
rs113242819 Chr.22: 26464590-26464590 on Build GRCh38
rs1042492421 Chr.22: 26464610-26464610 on Build GRCh38
rs751013277 Chr.22: 26464305-26464305 on Build GRCh38
rs757426392 Chr.22: 26464496-26464496 on Build GRCh38
rs781196776 Chr.22: 26464418-26464418 on Build GRCh38
rs773303744 Chr.22: 26464252-26464252 on Build GRCh38
rs1040644043 Chr.22: 26464666-26464666 on Build GRCh38
rs951352992 Chr.22: 26464372-26464372 on Build GRCh38
rs769041433 Chr.22: 26464514-26464514 on Build GRCh38
rs746418898 Chr.22: 26464363-26464363 on Build GRCh38
rs375526010 Chr.22: 26464453-26464453 on Build GRCh38
rs780435239 Chr.22: 26464432-26464432 on Build GRCh38
rs114685298 Chr.22: 26464570-26464570 on Build GRCh38
rs777875261 Chr.22: 26464348-26464348 on Build GRCh38
rs777094703 Chr.22: 26464360-26464360 on Build GRCh38
rs753983512 Chr.22: 26464589-26464589 on Build GRCh38
rs372499859 Chr.22: 26464338-26464338 on Build GRCh38
rs766233510 Chr.22: 26464399-26464399 on Build GRCh38
rs751823743 Chr.22: 26464412-26464412 on Build GRCh38
rs773460422 Chr.22: 26464661-26464661 on Build GRCh38
rs373523675 Chr.22: 26464433-26464433 on Build GRCh38
rs369053765 Chr.22: 26464498-26464498 on Build GRCh38
rs767548244 Chr.22: 26464314-26464314 on Build GRCh38
rs377095634 Chr.22: 26464519-26464519 on Build GRCh38
rs773931864 Chr.22: 26464386-26464386 on Build GRCh38
rs775881151 Chr.22: 26464370-26464370 on Build GRCh38
rs765186595 Chr.22: 26464562-26464562 on Build GRCh38
rs745384564 Chr.22: 26464636-26464636 on Build GRCh38
rs763228182 Chr.22: 26464378-26464378 on Build GRCh38
rs375587915 Chr.22: 26464260-26464260 on Build GRCh38
rs761480950 Chr.22: 26464532-26464534 on Build GRCh38
rs770266719 Chr.22: 26464428-26464428 on Build GRCh38
rs1054656108 Chr.22: 26464571-26464571 on Build GRCh38
rs548311034 Chr.22: 26464646-26464646 on Build GRCh38
rs748512719 Chr.22: 26464614-26464614 on Build GRCh38
rs144077166 Chr.22: 26464409-26464409 on Build GRCh38
rs766419128 Chr.22: 26464573-26464573 on Build GRCh38
rs770642869 Chr.22: 26464633-26464633 on Build GRCh38
rs756030696 Chr.22: 26464236-26464236 on Build GRCh38
rs1046054798 Chr.22: 26464308-26464308 on Build GRCh38
rs116769827 Chr.22: 26464569-26464569 on Build GRCh38
rs753787422 Chr.22: 26464483-26464483 on Build GRCh38
rs147435410 Chr.22: 26464234-26464234 on Build GRCh38
rs768468362 Chr.22: 26464277-26464279 on Build GRCh38
rs957749322 Chr.22: 26464269-26464269 on Build GRCh38
rs746693127 Chr.22: 26464619-26464619 on Build GRCh38
rs143965829 Chr.22: 26464672-26464672 on Build GRCh38
rs281865164 Chr.22: 26464657-26464658 on Build GRCh38
rs372828090 Chr.22: 26464515-26464515 on Build GRCh38
rs763743681 Chr.22: 26464385-26464385 on Build GRCh38
rs748136926 Chr.22: 26464274-26464274 on Build GRCh38
rs148662419 Chr.22: 26464420-26464420 on Build GRCh38
rs755952793 Chr.22: 26464334-26464334 on Build GRCh38
rs766628788 Chr.22: 26464675-26464675 on Build GRCh38
rs756839100 Chr.22: 26464310-26464310 on Build GRCh38
rs377326385 Chr.22: 26464654-26464654 on Build GRCh38
rs558884663 Chr.22: 26464506-26464506 on Build GRCh38
rs5761543 Chr.22: 26464322-26464322 on Build GRCh38
rs753613070 Chr.22: 26464404-26464404 on Build GRCh38
rs770221086 Chr.22: 26464369-26464369 on Build GRCh38
rs532646940 Chr.22: 26464547-26464547 on Build GRCh38
rs766330282 Chr.22: 26464482-26464482 on Build GRCh38
rs754066787 Chr.22: 26464245-26464245 on Build GRCh38
rs772783723 Chr.22: 26464465-26464465 on Build GRCh38
rs372027593 Chr.22: 26464235-26464235 on Build GRCh38
rs761403871 Chr.22: 26464390-26464390 on Build GRCh38
rs370007632 Chr.22: 26464393-26464393 on Build GRCh38
rs774114985 Chr.22: 26464447-26464447 on Build GRCh38
rs188300485 Chr.22: 26464616-26464616 on Build GRCh38
rs769658980 Chr.22: 26464637-26464637 on Build GRCh38
rs775195314 Chr.22: 26464566-26464566 on Build GRCh38
rs774979651 Chr.22: 26464300-26464300 on Build GRCh38
rs747080557 Chr.22: 26464353-26464353 on Build GRCh38
rs762613783 Chr.22: 26464301-26464301 on Build GRCh38
rs765478898 Chr.22: 26464529-26464529 on Build GRCh38
rs201043011 Chr.22: 26464398-26464398 on Build GRCh38
rs1019595949 Chr.22: 26464458-26464458 on Build GRCh38
rs779849991 Chr.22: 26464335-26464335 on Build GRCh38
rs777491163 Chr.22: 26464354-26464354 on Build GRCh38
rs375655372 Chr.22: 26464438-26464438 on Build GRCh38
rs763915803 Chr.22: 26464655-26464655 on Build GRCh38
rs778998363 Chr.22: 26464243-26464243 on Build GRCh38
rs749799140 Chr.22: 26464512-26464512 on Build GRCh38
rs752507059 Chr.22: 26464492-26464492 on Build GRCh38
rs758902895 Chr.22: 26464615-26464615 on Build GRCh38

View Full Product Details