The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_022337.2 | NP_071732.1 | 1 - 2 | 287 | 77 |
XM_017017455.1 | XP_016872944.1 | 1 - 2 | 325 | 77 | |
XM_017017456.1 | XP_016872945.1 | 1 - 2 | 325 | 77 | |
GenBank mRNA | AF136172.1 | - | 1 - 2 | 208 | 77 |
AF235022.1 | - | 1 - 2 | 251 | 77 | |
AL546159.3 | - | 1 - 2 | 287 | 77 | |
BC015808.1 | - | 1 - 2 | 253 | 77 | |
BT009842.1 | - | 1 - 2 | 204 | 77 |