The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_025239.3 | NP_079515.2 | 2 - 3 | 324 | 61 |
XM_005251600.3 | XP_005251657.1 | 2 - 3 | 431 | 61 | |
GenBank mRNA | AF329193.1 | - | 2 - 3 | 309 | 61 |
AF344424.1 | - | 2 - 3 | 324 | 61 | |
AY254343.1 | - | 1 - 2 | 51 | 61 | |
AY271901.1 | - | 1 - 2 | 51 | 61 | |
AY271902.1 | - | 1 - 2 | 51 | 61 | |
BC074766.2 | - | 2 - 3 | 85 | 61 | |
BC113678.1 | - | 2 - 3 | 87 | 61 | |
BC113680.1 | - | 2 - 3 | 87 | 61 | |
BC143920.1 | - | 2 - 3 | 87 | 61 | |
EF444806.1 | - | 2 - 3 | 171 | 61 |