The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_025243.3 | NP_079519.1 | 4 - 5 | 1261 | 99 |
XM_005246874.3 | XP_005246931.1 | 5 - 6 | 1424 | 99 | |
XM_011511931.2 | XP_011510233.1 | 4 - 5 | 1516 | 99 | |
XM_011511932.1 | XP_011510234.1 | 4 - 5 | 1338 | 99 | |
XM_011511933.1 | XP_011510235.1 | 4 - 5 | 1604 | 99 | |
XM_017005030.1 | XP_016860519.1 | 5 - 6 | 1606 | 99 | |
XM_017005031.1 | XP_016860520.1 | 5 - 6 | 1585 | 99 | |
XM_017005032.1 | XP_016860521.1 | 5 - 6 | 1466 | 99 | |
XM_017005033.1 | XP_016860522.1 | 5 - 6 | 1780 | 99 | |
XM_017005034.1 | XP_016860523.1 | 5 - 6 | 1710 | 99 | |
GenBank mRNA | AF271633.1 | - | 4 - 5 | 1215 | 99 |
AK301490.1 | - | 5 - 6 | 1427 | 99 | |
AK312464.1 | - | 4 - 5 | 1263 | 99 | |
BC032014.1 | - | 4 - 5 | 1216 | 99 |