The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_030761.4 | NP_110388.2 | 4 - 5 | 694 | 66 |
XM_011541597.2 | XP_011539899.1 | 4 - 5 | 2168 | 66 | |
XM_011541598.2 | XP_011539900.1 | 4 - 5 | 763 | 66 | |
GenBank mRNA | AB061675.1 | - | 4 - 5 | 595 | 66 |
AF316543.1 | - | 4 - 5 | 634 | 66 | |
AK296058.1 | - | 4 - 5 | 674 | 66 | |
AY009398.1 | - | 4 - 5 | 590 | 66 | |
AY358947.1 | - | 4 - 5 | 626 | 66 | |
BC057781.1 | - | 4 - 5 | 785 | 66 | |
BT020125.1 | - | 4 - 5 | 590 | 66 |