Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
AGAP3
Entrez Gene ID
116988
Gene Name
ArfGAP with GTPase domain, ankyrin repeat and PH domain 3
Gene Aliases
AGAP-3, CENTG3, CRAG, MRIP-1, cnt-g3
Chromosome Location
Chr.7: 151085831 - 151144436 on Build GRCh38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001281300.1 NP_001268229.1 14 - 15 1811 69
NM_031946.6 NP_114152.3 16 - 17 2592 69
XM_005249943.2 XP_005250000.1 16 - 17 2167 69
XM_011515780.1 XP_011514082.1 16 - 17 2204 69
XM_017011732.1 XP_016867221.1 14 - 15 1782 69
XM_017011733.1 XP_016867222.1 12 - 13 1473 69
XM_017011734.1 XP_016867223.1 14 - 15 1722 69
XM_017011735.1 XP_016867224.1 15 - 16 2202 69
GenBank mRNA AF359283.3 - 17 - 18 2167 69
AF413079.1 - 16 - 17 2167 69
AK027415.1 - 5 - 6 658 69
AK055393.1 - 14 - 15 1775 69
AL442089.1 - 3 - 4 2798 69
BC037543.1 - 6 - 7 798 69
BC048300.1 - 5 - 6 750 69

View Full Product Details