The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_001202513.1 | NP_001189442.1 | 3 - 4 | 492 | 60 |
NM_002357.3 | NP_002348.1 | 3 - 4 | 492 | 60 | |
GenBank mRNA | AK312734.1 | - | 3 - 4 | 425 | 60 |
BC069377.1 | - | 3 - 4 | 286 | 60 | |
BC069433.1 | - | 3 - 4 | 286 | 60 | |
BC098396.1 | - | 3 - 4 | 462 | 60 | |
BC113531.1 | - | 3 - 4 | 345 | 60 | |
BC117260.1 | - | 3 - 4 | 345 | 60 | |
BC143832.1 | - | 3 - 4 | 345 | 60 | |
CR536495.1 | - | 3 - 4 | 202 | 60 | |
CR541692.1 | - | 3 - 4 | 202 | 60 | |
DC347823.1 | - | 3 - 4 | 493 | 60 | |
L06895.1 | - | 3 - 4 | 349 | 60 |