The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_005188.3 | NP_005179.2 | 12 - 13 | 2178 | 64 |
XM_017018511.1 | XP_016874000.1 | 11 - 12 | 1613 | 64 | |
GenBank mRNA | BC132733.1 | - | 12 - 13 | 2107 | 64 |
BC136463.1 | - | 12 - 13 | 2107 | 64 | |
X57110.1 | - | 12 - 13 | 2184 | 64 |