The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_001190737.1 | NP_001177666.1 | 5 - 6 | 1345 | 130 |
NM_001190738.1 | NP_001177667.1 | 5 - 6 | 1240 | 130 | |
NM_001282787.1 | NP_001269716.1 | 4 - 5 | 392 | 130 | |
NM_005596.3 | NP_005587.2 | 5 - 6 | 1345 | 130 | |
XM_005251467.2 | XP_005251524.1 | 5 - 6 | 1438 | 130 | |
XM_005251468.4 | XP_005251525.1 | 5 - 6 | 911 | 130 | |
XM_005251469.2 | XP_005251526.1 | 5 - 6 | 1438 | 130 | |
XM_005251470.2 | XP_005251527.1 | 5 - 6 | 1438 | 130 | |
XM_005251471.3 | XP_005251528.1 | 4 - 5 | 2273 | 130 | |
XM_006716773.2 | XP_006716836.1 | 5 - 6 | 1438 | 130 | |
XM_006716774.2 | XP_006716837.1 | 5 - 6 | 1438 | 130 | |
XM_006716775.2 | XP_006716838.1 | 5 - 6 | 1438 | 130 | |
XM_017014739.1 | XP_016870228.1 | 5 - 6 | 1438 | 130 | |
XM_017014740.1 | XP_016870229.1 | 5 - 6 | 1256 | 130 | |
XM_017014741.1 | XP_016870230.1 | 5 - 6 | 1376 | 130 | |
XM_017014742.1 | XP_016870231.1 | 4 - 5 | 2799 | 130 | |
GenBank mRNA | AK131233.1 | - | 5 - 6 | 1240 | 130 |
AK290603.1 | - | 5 - 6 | 958 | 130 | |
AK299605.1 | - | 5 - 6 | 1208 | 130 | |
BC001283.1 | - | 5 - 6 | 968 | 130 | |
BP372854.1 | - | 4 - 5 | 392 | 130 | |
BT007266.1 | - | 5 - 6 | 810 | 130 | |
BX648416.1 | - | 4 - 5 | 389 | 130 | |
KU178232.1 | - | 5 - 6 | 810 | 130 | |
KU178233.1 | - | 6 - 7 | 888 | 130 | |
U85193.1 | - | 5 - 6 | 1019 | 130 |