The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_001130042.1 | NP_001123514.1 | 4 - 5 | 765 | 72 |
NM_001130043.1 | NP_001123515.1 | 3 - 4 | 726 | 72 | |
NM_001889.3 | NP_001880.2 | 3 - 4 | 726 | 72 | |
XM_011540747.1 | XP_011539049.1 | 3 - 4 | 345 | 72 | |
XM_017000367.1 | XP_016855856.1 | 4 - 5 | 400 | 72 | |
GenBank mRNA | AB209714.1 | - | 4 - 5 | 397 | 72 |
AK128794.1 | - | 3 - 4 | 331 | 72 | |
AK223150.1 | - | 3 - 4 | 358 | 72 | |
AK223201.1 | - | 3 - 4 | 358 | 72 | |
AK314813.1 | - | 3 - 4 | 366 | 72 | |
BC039578.1 | - | 3 - 4 | 348 | 72 | |
BC070058.1 | - | 4 - 5 | 407 | 72 | |
DA256305.1 | - | 3 - 4 | 410 | 72 | |
L13278.1 | - | 2 - 3 | 270 | 72 |