The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_001080955.2 | NP_001074424.1 | 5 - 6 | 363 | 79 |
NM_001080956.2 | NP_001074425.1 | 5 - 6 | 558 | 79 | |
NM_001289037.1 | NP_001275966.1 | 4 - 5 | 322 | 79 | |
NM_001289038.1 | NP_001275967.1 | 3 - 4 | 276 | 79 | |
NM_001289040.1 | NP_001275969.1 | 5 - 6 | 348 | 79 | |
NM_001289041.1 | NP_001275970.1 | 5 - 6 | 558 | 79 | |
NM_001317158.1 | NP_001304087.1 | 2 - 3 | 778 | 79 | |
NM_001317160.1 | NP_001304089.1 | 5 - 6 | 544 | 79 | |
GenBank mRNA | AJ303119.1 | - | 1 - 2 | 86 | 79 |
AK314570.1 | - | 5 - 6 | 405 | 79 | |
BX537397.1 | - | 2 - 3 | 3747 | 79 | |
DA426770.1 | - | 5 - 6 | 405 | 79 | |
DA549753.1 | - | 5 - 6 | 509 | 79 | |
DA675185.1 | - | 5 - 6 | 357 | 79 | |
DA717476.1 | - | 5 - 6 | 398 | 79 | |
U81992.1 | - | 2 - 3 | 170 | 79 |