The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_004327.3 | NP_004318.3 | 16 - 17 | 3611 | 81 |
GenBank mRNA | AB209991.1 | - | 16 - 17 | 3360 | 81 |
AF487522.1 | - | 1 - 2 | 72 | 81 | |
AK303241.1 | - | 16 - 17 | 2045 | 81 | |
BC066122.1 | - | 16 - 17 | 3442 | 81 | |
BQ422888.1 | - | 7 - 8 | 737 | 81 | |
X02596.1 | - | 16 - 17 | 3503 | 81 |