The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_001037328.2 | NP_001032405.1 | 8 - 9 | 1231 | 54 |
NM_001319134.1 | NP_001306063.1 | 7 - 8 | 1108 | 54 | |
NM_001319135.1 | NP_001306064.1 | 9 - 10 | 1290 | 54 | |
NM_001322927.1 | NP_001309856.1 | 6 - 7 | 1033 | 54 | |
NM_001322928.1 | NP_001309857.1 | 9 - 10 | 1377 | 54 | |
NM_001322929.1 | NP_001309858.1 | 9 - 10 | 1502 | 54 | |
NM_001322930.1 | NP_001309859.1 | 7 - 8 | 1304 | 54 | |
NM_001322931.1 | NP_001309860.1 | 10 - 11 | 1482 | 54 | |
NM_001322932.1 | NP_001309861.1 | 7 - 8 | 1322 | 54 | |
NM_001322933.1 | NP_001309862.1 | 8 - 9 | 1397 | 54 | |
NM_004602.3 | NP_004593.2 | 8 - 9 | 1213 | 54 | |
NM_017452.3 | NP_059346.2 | 7 - 8 | 1090 | 54 | |
NM_017453.3 | NP_059347.2 | 8 - 9 | 1379 | 54 | |
NM_017454.3 | NP_059348.2 | 6 - 7 | 1015 | 54 | |
XM_005260527.1 | XP_005260584.1 | 8 - 9 | 1280 | 54 | |
XM_006723865.1 | XP_006723928.1 | 9 - 10 | 1517 | 54 | |
XM_011529015.1 | XP_011527317.1 | 10 - 11 | 1630 | 54 | |
XM_017028028.1 | XP_016883517.1 | 9 - 10 | 1356 | 54 | |
GenBank mRNA | AB209561.1 | - | 8 - 9 | 1244 | 54 |
AF061938.1 | - | 7 - 8 | 1084 | 54 | |
AF061939.1 | - | 8 - 9 | 1373 | 54 | |
AF061940.1 | - | 6 - 7 | 1009 | 54 | |
AF061941.1 | - | 8 - 9 | 1216 | 54 | |
AJ132258.1 | - | 5 - 6 | 757 | 54 | |
AK025519.1 | - | 7 - 8 | 1052 | 54 | |
AK091405.1 | - | 4 - 5 | 569 | 54 | |
AK291487.1 | - | 8 - 9 | 1294 | 54 | |
AK292859.1 | - | 7 - 8 | 875 | 54 | |
AL136601.2 | - | 7 - 8 | 1073 | 54 | |
AY529074.1 | - | 7 - 8 | 926 | 54 | |
AY546099.1 | - | 7 - 8 | 1102 | 54 | |
BC050432.1 | - | 7 - 8 | 1117 | 54 | |
BC095397.1 | - | 7 - 8 | 1062 | 54 |