The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_004562.2 | NP_004553.2 | 8 - 9 | 1069 | 74 |
NM_013987.2 | NP_054642.2 | 7 - 8 | 985 | 74 | |
NM_013988.2 | NP_054643.2 | 5 - 6 | 622 | 74 | |
XM_011535863.1 | XP_011534165.1 | 8 - 9 | 1066 | 74 | |
XM_017010908.1 | XP_016866397.1 | 8 - 9 | 1983 | 74 | |
XM_017010909.1 | XP_016866398.1 | 7 - 8 | 863 | 74 | |
GenBank mRNA | AB009973.1 | - | 8 - 9 | 1036 | 74 |
AB245403.1 | - | 9 - 10 | 1027 | 74 | |
AF381282.1 | - | 6 - 7 | 694 | 74 | |
AF381284.1 | - | 6 - 7 | 813 | 74 | |
AF381286.1 | - | 1 - 2 | 71 | 74 | |
AK292590.1 | - | 8 - 9 | 1030 | 74 | |
AK294684.1 | - | 3 - 4 | 275 | 74 | |
BC022014.2 | - | 9 - 10 | 1090 | 74 | |
GU345837.1 | - | 7 - 8 | 788 | 74 | |
GU345838.1 | - | 6 - 7 | 830 | 74 | |
GU345839.1 | - | 7 - 8 | 868 | 74 | |
GU357501.1 | - | 5 - 6 | 425 | 74 | |
GU361469.1 | - | 8 - 9 | 1049 | 74 | |
GU361470.1 | - | 8 - 9 | 1050 | 74 | |
KC357595.1 | - | 10 - 11 | 1147 | 74 | |
KC774171.1 | - | 6 - 7 | 802 | 74 | |
KJ534911.1 | - | 7 - 8 | 935 | 74 | |
KJ534912.1 | - | 7 - 8 | 935 | 74 | |
KU178240.1 | - | 8 - 9 | 992 | 74 | |
KU178241.1 | - | - | 935 | 74 | |
KU178242.1 | - | - | 932 | 74 |