The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_001320591.1 | NP_001307520.1 | 14 - 15 | 2044 | 126 |
NM_001320592.1 | NP_001307521.1 | 13 - 14 | 2107 | 126 | |
NM_001320593.1 | NP_001307522.1 | 13 - 14 | 2103 | 126 | |
NM_001320594.1 | NP_001307523.1 | 14 - 15 | 2177 | 126 | |
NM_016824.4 | NP_058432.1 | 13 - 14 | 2123 | 126 | |
GenBank mRNA | AB209810.1 | - | 13 - 14 | 1886 | 126 |
AK223270.1 | - | 14 - 15 | 1713 | 126 | |
AK291488.1 | - | 13 - 14 | 1939 | 126 | |
BC043143.1 | - | 13 - 14 | 1848 | 126 | |
BC057285.1 | - | 13 - 14 | 1833 | 126 | |
BC062559.1 | - | 13 - 14 | 2096 | 126 | |
BX647442.1 | - | 4 - 5 | 1584 | 126 | |
EF553523.1 | - | 14 - 15 | 2045 | 126 |