The assay primers and probes lie within a single exon. This assay will detect gDNA.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_023016.3 | NP_075392.2 | 1 - 1 | 1866 | 146 |
GenBank mRNA | AK023346.1 | - | 1 - 1 | 146 | 146 |
AK025523.1 | - | 1 - 1 | 1835 | 146 | |
BC036652.1 | - | 1 - 1 | 1890 | 146 |