Gene Symbol | Transcript Accession | Exon Location | Assay Location | Protein ID |
---|---|---|---|---|
FOXP3 | NM_001114377.1 | NP_001107849.1 | ||
NM_014009.3 | NP_054728.2 | |||
XM_006724533.2 | XP_006724596.2 | |||
XM_011543916.2 | XP_011542218.1 | |||
XM_017029565.1 | XP_016885054.1 | |||
XM_017029566.1 | XP_016885055.1 | |||
XM_017029567.1 | XP_016885056.1 | |||
AF277993.1 | AAG53607.1 | |||
AJ005891.1 | CAA06748.1 | |||
AK292052.1 | ||||
AK299988.1 | ||||
BC113401.1 | ||||
BC113403.1 | ||||
BC143785.1 | ||||
BC143786.1 | ||||
DQ010327.1 | AAY27088.1 | |||
EF534714.1 | ||||
EU855812.1 |
Target Variation |
Location | CNV Subtype |
Genes |
---|---|---|---|
nsv471602 | ChrX:49205142 - 49320261 on Build GRCh38 | Gain | FOXP3 GAGE10 CCDC22 PPP1R3F CACNA1F |
nsv528292 | ChrX:48433286 - 49274368 on Build GRCh38 | Gain | WDR45 MAGIX EBP SUV39H1 LOC101927635 SLC35A2 GLOD5 PCSK1N PLP2 PRAF2 SYP-AS1 ERAS GATA1 CCDC22 SLC38A5 OTUD5 FOXP3 PQBP1 CCDC120 TIMM17B PPP1R3F KCND1 CACNA1F GPKOW TBC1D25 PRICKLE3 FTSJ1 RBM3 WAS TFE3 PIM2 PORCN GRIPAP1 WDR13 HDAC6 SYP |
esv32909 | ChrX:48003374 - 49285646 on Build GRCh38 | Gain+Loss | SSX9 WDR45 ZNF630-AS1 MAGIX EBP ZNF630 SUV39H1 SPACA5B LOC101927635 SLC35A2 GLOD5 PCSK1N PLP2 SSX6 PRAF2 SYP-AS1 ERAS GATA1 CCDC22 SSX1 SLC38A5 OTUD5 SSX3 SSX4B FOXP3 SSX5 PQBP1 CCDC120 TIMM17B PPP1R3F KCND1 CACNA1F GPKOW TBC1D25 PRICKLE3 FTSJ1 RBM3 SPACA5 WAS TFE3 PIM2 ZNF182 PORCN GRIPAP1 SSX4 WDR13 HDAC6 SYP |
nsv469734 | ChrX:49205141 - 49320261 on Build GRCh38 | Gain+Loss | FOXP3 GAGE10 CCDC22 PPP1R3F CACNA1F |
nsv6895 | ChrX:49227319 - 49262690 on Build GRCh38 | Insertion | FOXP3 CCDC22 CACNA1F |