Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
FASTKD1
Entrez Gene ID
79675
Gene Name
FAST kinase domains 1
Gene Aliases
-
Chromosome Location
Chr.2: 169527879 - 169573921 on Build GRCh38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001281476.2 NP_001268405.1 8 - 9 2093 74
NM_001322046.1 NP_001308975.1 8 - 9 2766 74
NM_001322048.1 NP_001308977.1 7 - 8 2021 74
NM_001322049.1 NP_001308978.1 7 - 8 2024 74
NM_024622.5 NP_078898.3 8 - 9 2093 74
NR_104020.2 8 - 9 2093 74
XM_006712750.3 XP_006712813.1 7 - 8 2019 74
XM_006712751.3 XP_006712814.1 4 - 5 2104 74
XM_017004911.1 XP_016860400.1 8 - 9 3406 74
XM_017004912.1 XP_016860401.1 4 - 5 2104 74
GenBank mRNA AB058703.1 - 6 - 7 1275 74
AK025554.1 - 5 - 6 1205 74
AK055892.1 - 8 - 9 2004 74
AK074302.1 - 8 - 9 1984 74
AK225293.1 - 5 - 6 1205 74
AL832058.1 - 8 - 9 1976 74

View Full Product Details