Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.1: 155235319-155235815 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
GBA
Entrez Gene ID
2629
Gene Name
glucosylceramidase beta
Gene Aliases
GBA1, GCB, GLUC
UniGene
Hs.282997
Gene Chromosome Location
Chr.1: 155234448-155244862 on Build GRCh38
Transcripts
NM_000157.3 NM_001005741.2 NM_001005742.2 NM_001171811.1 NM_001171812.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
4463116 c.1277C>T p.P426L Chr.1: 155235792-155235792 on Build GRCh38
4022961 c.1267G>T p.A423S Chr.1: 155235802-155235802 on Build GRCh38
463092 c.1367T>A p.F456Y Chr.1: 155235702-155235702 on Build GRCh38
3474893 c.1319C>T p.P440L Chr.1: 155235750-155235750 on Build GRCh38
5992377 c.1309G>T p.V437F Chr.1: 155235760-155235760 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs426516 Chr.1: 155235412-155235412 on Build GRCh38
rs78802049 Chr.1: 155235725-155235725 on Build GRCh38
rs772140702 Chr.1: 155235767-155235767 on Build GRCh38
rs79185870 Chr.1: 155235701-155235701 on Build GRCh38
rs778798290 Chr.1: 155235764-155235764 on Build GRCh38
rs75385858 Chr.1: 155235765-155235765 on Build GRCh38
rs202221385 Chr.1: 155235732-155235732 on Build GRCh38
rs761062637 Chr.1: 155235659-155235659 on Build GRCh38
rs75564605 Chr.1: 155235747-155235747 on Build GRCh38
rs994723035 Chr.1: 155235792-155235792 on Build GRCh38
rs750193229 Chr.1: 155235722-155235722 on Build GRCh38
rs80020805 Chr.1: 155235704-155235704 on Build GRCh38
rs775664680 Chr.1: 155235361-155235361 on Build GRCh38
rs906266554 Chr.1: 155235746-155235746 on Build GRCh38
rs75090908 Chr.1: 155235755-155235755 on Build GRCh38
rs80356769 Chr.1: 155235772-155235772 on Build GRCh38
rs75243000 Chr.1: 155235762-155235762 on Build GRCh38
rs754798497 Chr.1: 155235737-155235737 on Build GRCh38
rs552762192 Chr.1: 155235478-155235478 on Build GRCh38
rs1800473 Chr.1: 155235540-155235540 on Build GRCh38
rs1064651 Chr.1: 155235727-155235727 on Build GRCh38
rs774554048 Chr.1: 155235347-155235347 on Build GRCh38
rs758671508 Chr.1: 155235349-155235350 on Build GRCh38
rs772349239 Chr.1: 155235358-155235358 on Build GRCh38
rs149171124 Chr.1: 155235790-155235790 on Build GRCh38
rs775661835 Chr.1: 155235768-155235768 on Build GRCh38
rs764486586 Chr.1: 155235661-155235661 on Build GRCh38
rs919900295 Chr.1: 155235448-155235448 on Build GRCh38
rs1011861500 Chr.1: 155235654-155235654 on Build GRCh38
rs542894715 Chr.1: 155235626-155235626 on Build GRCh38
rs77738682 Chr.1: 155235777-155235777 on Build GRCh38
rs757930613 Chr.1: 155235723-155235723 on Build GRCh38
rs944242689 Chr.1: 155235512-155235512 on Build GRCh38
rs77369218 Chr.1: 155235726-155235726 on Build GRCh38
rs781189218 Chr.1: 155235751-155235751 on Build GRCh38
rs2974924 Chr.1: 155235379-155235379 on Build GRCh38
rs121908295 Chr.1: 155235708-155235708 on Build GRCh38
rs189413365 Chr.1: 155235571-155235571 on Build GRCh38
rs1053388016 Chr.1: 155235612-155235612 on Build GRCh38
rs74598136 Chr.1: 155235750-155235750 on Build GRCh38
rs577592360 Chr.1: 155235479-155235479 on Build GRCh38
rs78346899 Chr.1: 155235716-155235716 on Build GRCh38
rs572918635 Chr.1: 155235668-155235668 on Build GRCh38
rs79032178 Chr.1: 155235758-155235758 on Build GRCh38
rs747284798 Chr.1: 155235769-155235769 on Build GRCh38
rs548372618 Chr.1: 155235389-155235389 on Build GRCh38
rs762367232 Chr.1: 155235672-155235672 on Build GRCh38
rs569191841 Chr.1: 155235344-155235344 on Build GRCh38
rs766764173 Chr.1: 155235632-155235634 on Build GRCh38
rs80356768 Chr.1: 155235752-155235806 on Build GRCh38
rs931386996 Chr.1: 155235453-155235453 on Build GRCh38
rs183510604 Chr.1: 155235409-155235409 on Build GRCh38
rs121908310 Chr.1: 155235760-155235760 on Build GRCh38
rs201499639 Chr.1: 155235791-155235791 on Build GRCh38
rs12752133 Chr.1: 155235587-155235587 on Build GRCh38
rs554812953 Chr.1: 155235660-155235660 on Build GRCh38
rs935574359 Chr.1: 155235607-155235607 on Build GRCh38
rs777049786 Chr.1: 155235776-155235776 on Build GRCh38
rs77035024 Chr.1: 155235719-155235719 on Build GRCh38
rs76910485 Chr.1: 155235780-155235780 on Build GRCh38
rs1026993108 Chr.1: 155235794-155235794 on Build GRCh38
rs1057519357 Chr.1: 155235792-155235792 on Build GRCh38
rs74752878 Chr.1: 155235699-155235699 on Build GRCh38
rs377666098 Chr.1: 155235343-155235343 on Build GRCh38

View Full Product Details