Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.6: 144007394-144007922 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
HYMAI
Entrez Gene ID
57061
Gene Name
hydatidiform mole associated and imprinted (non-protein coding)
Gene Aliases
NCRNA00020
UniGene
Hs.657760
Gene Chromosome Location
Chr.6: 144004916-144008730 on Build GRCh38
Transcripts
NR_002768.2 NM_001080951.2 NM_001080952.2 NM_001080953.2 NM_001080954.2 NM_001080955.2 NM_001080956.2 NM_001289037.1 NM_001289038.1 NM_001289039.1 NM_001289040.1 NM_001289041.1 NM_001289042.1 NM_001289043.1 NM_001289044.1 NM_001289045.1 NM_001289046.1 NM_001289047.1 NM_001289048.1 NM_001289049.1 NM_001317159.1 NM_001317160.1 NM_001317161.1 NM_001317162.1 NM_006718.4

Gene Symbol
PLAGL1
Entrez Gene ID
5325
Gene Name
PLAG1 like zinc finger 1
Gene Aliases
LOT1, ZAC, ZAC1
UniGene
Hs.743225
Gene Chromosome Location
Chr.6: 143940300-144064599 on Build GRCh38
Transcripts
NR_002768.2 NM_001080951.2 NM_001080952.2 NM_001080953.2 NM_001080954.2 NM_001080955.2 NM_001080956.2 NM_001289037.1 NM_001289038.1 NM_001289039.1 NM_001289040.1 NM_001289041.1 NM_001289042.1 NM_001289043.1 NM_001289044.1 NM_001289045.1 NM_001289046.1 NM_001289047.1 NM_001289048.1 NM_001289049.1 NM_001317159.1 NM_001317160.1 NM_001317161.1 NM_001317162.1 NM_006718.4

SNP Detail

SNP ID Chromosome Location
rs921554812 Chr.6: 144007593-144007593 on Build GRCh38
rs913957075 Chr.6: 144007688-144007688 on Build GRCh38
rs1017067025 Chr.6: 144007886-144007886 on Build GRCh38
rs573784560 Chr.6: 144007844-144007844 on Build GRCh38
rs776020539 Chr.6: 144007451-144007451 on Build GRCh38
rs73006277 Chr.6: 144007534-144007534 on Build GRCh38
rs934131164 Chr.6: 144007605-144007605 on Build GRCh38
rs926950535 Chr.6: 144007733-144007733 on Build GRCh38
rs758235766 Chr.6: 144007702-144007705 on Build GRCh38
rs938055905 Chr.6: 144007776-144007776 on Build GRCh38
rs1018242536 Chr.6: 144007588-144007588 on Build GRCh38
rs2281476 Chr.6: 144007831-144007831 on Build GRCh38
rs938615426 Chr.6: 144007771-144007771 on Build GRCh38
rs1014067670 Chr.6: 144007806-144007807 on Build GRCh38
rs998128998 Chr.6: 144007861-144007861 on Build GRCh38
rs1005677401 Chr.6: 144007873-144007873 on Build GRCh38
rs1006948675 Chr.6: 144007845-144007845 on Build GRCh38
rs966029932 Chr.6: 144007619-144007619 on Build GRCh38
rs951123783 Chr.6: 144007535-144007535 on Build GRCh38
rs568685211 Chr.6: 144007434-144007434 on Build GRCh38
rs891701520 Chr.6: 144007804-144007804 on Build GRCh38
rs1034305112 Chr.6: 144007446-144007446 on Build GRCh38
rs903845751 Chr.6: 144007766-144007766 on Build GRCh38
rs762708243 Chr.6: 144007787-144007787 on Build GRCh38
rs980182181 Chr.6: 144007703-144007703 on Build GRCh38
rs1025886660 Chr.6: 144007869-144007869 on Build GRCh38
rs938577492 Chr.6: 144007485-144007485 on Build GRCh38
rs886816035 Chr.6: 144007816-144007816 on Build GRCh38
rs148450783 Chr.6: 144007568-144007568 on Build GRCh38
rs951630420 Chr.6: 144007870-144007870 on Build GRCh38
rs868867877 Chr.6: 144007843-144007843 on Build GRCh38
rs1040130654 Chr.6: 144007848-144007848 on Build GRCh38
rs909006612 Chr.6: 144007663-144007663 on Build GRCh38
rs951578476 Chr.6: 144007656-144007656 on Build GRCh38
rs369535317 Chr.6: 144007887-144007887 on Build GRCh38
rs901199666 Chr.6: 144007850-144007850 on Build GRCh38
rs759023237 Chr.6: 144007493-144007493 on Build GRCh38
rs979474678 Chr.6: 144007884-144007884 on Build GRCh38
rs986894011 Chr.6: 144007865-144007865 on Build GRCh38
rs539805658 Chr.6: 144007512-144007512 on Build GRCh38
rs984247423 Chr.6: 144007657-144007657 on Build GRCh38
rs144569241 Chr.6: 144007433-144007433 on Build GRCh38
rs904401155 Chr.6: 144007436-144007436 on Build GRCh38
rs1025834254 Chr.6: 144007647-144007647 on Build GRCh38
rs976061286 Chr.6: 144007590-144007590 on Build GRCh38
rs941749221 Chr.6: 144007702-144007702 on Build GRCh38
rs1000518458 Chr.6: 144007441-144007441 on Build GRCh38
rs535732467 Chr.6: 144007443-144007443 on Build GRCh38
rs557631847 Chr.6: 144007487-144007487 on Build GRCh38
rs982409964 Chr.6: 144007559-144007559 on Build GRCh38
rs865946168 Chr.6: 144007579-144007579 on Build GRCh38
rs569490803 Chr.6: 144007498-144007498 on Build GRCh38
rs945895581 Chr.6: 144007806-144007806 on Build GRCh38
rs1014111088 Chr.6: 144007524-144007524 on Build GRCh38
rs540346745 Chr.6: 144007612-144007612 on Build GRCh38
rs1021745510 Chr.6: 144007578-144007578 on Build GRCh38
rs975125950 Chr.6: 144007851-144007851 on Build GRCh38
rs977009569 Chr.6: 144007634-144007634 on Build GRCh38
rs1007307325 Chr.6: 144007587-144007587 on Build GRCh38
rs894410480 Chr.6: 144007781-144007781 on Build GRCh38
rs1056098160 Chr.6: 144007777-144007777 on Build GRCh38

View Full Product Details