Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
AMDHD1
Entrez Gene ID
144193
Gene Name
amidohydrolase domain containing 1
Gene Aliases
HMFT1272
Chromosome Location
Chr.12: 95943293 - 95968592 on Build GRCh38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_152435.2 NP_689648.2 6 - 7 1050 92
GenBank mRNA AB075878.1 - 6 - 7 1005 92
AK054617.1 - 6 - 7 1050 92
AK122802.1 - 6 - 7 1017 92
AK290828.1 - 6 - 7 1005 92
AL833144.1 - 5 - 6 758 92
BC029146.1 - 6 - 7 986 92

View Full Product Details