Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.7: 65979649-65980150 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
GUSB
Entrez Gene ID
2990
Gene Name
glucuronidase beta
Gene Aliases
BG, MPS7
UniGene
Hs.255230
Gene Chromosome Location
Chr.7: 65960684-65982314 on Build GRCh38
Transcripts
NM_000181.3 NM_001284290.1 NM_001293104.1 NM_001293105.1 NR_120531.1 XM_005250297.3 XM_011516113.1 XM_011516114.1 XM_017012091.1 XM_017012092.1 XM_017012093.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
1623045 c.482C>G p.P161R Chr.7: 65979826-65979826 on Build GRCh38
5710642 c.491C>T p.S164F Chr.7: 65979817-65979817 on Build GRCh38
1292533 c.397T>G p.W133G Chr.7: 65979911-65979911 on Build GRCh38
291738 c.534_535insC p.T179fs*13 Chr.7: 65979773-65979774 on Build GRCh38
6867332 c.432G>T p.G144G Chr.7: 65979876-65979876 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs752674900 Chr.7: 65979798-65979798 on Build GRCh38
rs991057458 Chr.7: 65980098-65980098 on Build GRCh38
rs771063329 Chr.7: 65979914-65979914 on Build GRCh38
rs781460305 Chr.7: 65979690-65979690 on Build GRCh38
rs900224467 Chr.7: 65980012-65980012 on Build GRCh38
rs779475930 Chr.7: 65979789-65979789 on Build GRCh38
rs1028594511 Chr.7: 65979808-65979808 on Build GRCh38
rs149606212 Chr.7: 65979854-65979854 on Build GRCh38
rs771137253 Chr.7: 65979815-65979815 on Build GRCh38
rs773134072 Chr.7: 65979810-65979810 on Build GRCh38
rs944546037 Chr.7: 65979972-65979972 on Build GRCh38
rs121918177 Chr.7: 65979866-65979866 on Build GRCh38
rs587779400 Chr.7: 65979778-65979778 on Build GRCh38
rs780835314 Chr.7: 65979892-65979892 on Build GRCh38
rs527730129 Chr.7: 65979719-65979719 on Build GRCh38
rs969869681 Chr.7: 65979877-65979877 on Build GRCh38
rs907575462 Chr.7: 65979965-65979965 on Build GRCh38
rs184274015 Chr.7: 65979976-65979976 on Build GRCh38
rs569078357 Chr.7: 65980011-65980011 on Build GRCh38
rs756936015 Chr.7: 65979694-65979694 on Build GRCh38
rs1047823381 Chr.7: 65979730-65979730 on Build GRCh38
rs768029322 Chr.7: 65979756-65979756 on Build GRCh38
rs1010010852 Chr.7: 65979812-65979812 on Build GRCh38
rs756417262 Chr.7: 65979763-65979763 on Build GRCh38
rs959520969 Chr.7: 65980093-65980094 on Build GRCh38
rs970090847 Chr.7: 65980116-65980116 on Build GRCh38
rs983438482 Chr.7: 65980118-65980118 on Build GRCh38
rs996983482 Chr.7: 65979790-65979790 on Build GRCh38
rs539767109 Chr.7: 65980097-65980097 on Build GRCh38
rs928572623 Chr.7: 65979898-65979898 on Build GRCh38
rs938631960 Chr.7: 65979905-65979905 on Build GRCh38
rs776287966 Chr.7: 65979778-65979781 on Build GRCh38
rs774576359 Chr.7: 65979921-65979921 on Build GRCh38
rs1021674783 Chr.7: 65980114-65980114 on Build GRCh38
rs781205550 Chr.7: 65979800-65979800 on Build GRCh38
rs765594686 Chr.7: 65979930-65979931 on Build GRCh38
rs759668020 Chr.7: 65979930-65979930 on Build GRCh38
rs767952315 Chr.7: 65979990-65979990 on Build GRCh38
rs773510214 Chr.7: 65979726-65979726 on Build GRCh38
rs754898172 Chr.7: 65979685-65979685 on Build GRCh38
rs754452177 Chr.7: 65979888-65979888 on Build GRCh38
rs376196268 Chr.7: 65979931-65979931 on Build GRCh38
rs58487691 Chr.7: 65979912-65979912 on Build GRCh38
rs748464871 Chr.7: 65979691-65979691 on Build GRCh38
rs1039921307 Chr.7: 65979770-65979770 on Build GRCh38
rs754692766 Chr.7: 65980076-65980076 on Build GRCh38
rs369537067 Chr.7: 65979794-65979794 on Build GRCh38
rs576100461 Chr.7: 65979676-65979676 on Build GRCh38
rs769769379 Chr.7: 65979809-65979809 on Build GRCh38
rs952081678 Chr.7: 65980113-65980114 on Build GRCh38
rs749073290 Chr.7: 65979901-65979901 on Build GRCh38
rs763253448 Chr.7: 65979738-65979738 on Build GRCh38
rs3061581 Chr.7: 65980093-65980094 on Build GRCh38
rs148364905 Chr.7: 65979861-65979861 on Build GRCh38
rs775063026 Chr.7: 65979699-65979699 on Build GRCh38
rs778102597 Chr.7: 65979775-65979775 on Build GRCh38
rs141441283 Chr.7: 65979897-65979897 on Build GRCh38
rs1021669842 Chr.7: 65979875-65979875 on Build GRCh38
rs548022773 Chr.7: 65980031-65980031 on Build GRCh38
rs867506320 Chr.7: 65980077-65980077 on Build GRCh38
rs1027860249 Chr.7: 65980101-65980101 on Build GRCh38
rs764486500 Chr.7: 65979858-65979858 on Build GRCh38
rs1032117020 Chr.7: 65979856-65979856 on Build GRCh38
rs920626481 Chr.7: 65979874-65979874 on Build GRCh38
rs891792789 Chr.7: 65980072-65980072 on Build GRCh38
rs753065037 Chr.7: 65979760-65979760 on Build GRCh38
rs768612200 Chr.7: 65979711-65979711 on Build GRCh38
rs746475728 Chr.7: 65979797-65979797 on Build GRCh38
rs376583332 Chr.7: 65979696-65979696 on Build GRCh38
rs906910887 Chr.7: 65980048-65980048 on Build GRCh38
rs113856819 Chr.7: 65980045-65980045 on Build GRCh38
rs11559281 Chr.7: 65979776-65979776 on Build GRCh38
rs786205671 Chr.7: 65979910-65979910 on Build GRCh38
rs901341646 Chr.7: 65979779-65979779 on Build GRCh38
rs746956912 Chr.7: 65979706-65979706 on Build GRCh38
rs759593913 Chr.7: 65979816-65979816 on Build GRCh38
rs751638113 Chr.7: 65979748-65979748 on Build GRCh38
rs1047796065 Chr.7: 65979700-65979700 on Build GRCh38
rs761222082 Chr.7: 65979940-65979940 on Build GRCh38
rs74430256 Chr.7: 65979747-65979747 on Build GRCh38
rs990678956 Chr.7: 65979870-65979870 on Build GRCh38
rs138435796 Chr.7: 65979768-65979768 on Build GRCh38
rs747969208 Chr.7: 65979806-65979806 on Build GRCh38
rs144397476 Chr.7: 65979855-65979855 on Build GRCh38
rs375740757 Chr.7: 65979879-65979879 on Build GRCh38
rs907946113 Chr.7: 65979964-65979964 on Build GRCh38
rs138955514 Chr.7: 65980032-65980032 on Build GRCh38
rs756893746 Chr.7: 65980113-65980113 on Build GRCh38
rs142873837 Chr.7: 65980063-65980063 on Build GRCh38
rs956058239 Chr.7: 65979864-65979864 on Build GRCh38
rs121918181 Chr.7: 65979782-65979782 on Build GRCh38
rs74745052 Chr.7: 65980030-65980030 on Build GRCh38
rs776586594 Chr.7: 65979713-65979713 on Build GRCh38
rs372648212 Chr.7: 65979686-65979686 on Build GRCh38
rs759839844 Chr.7: 65979922-65979957 on Build GRCh38
rs533581698 Chr.7: 65980042-65980042 on Build GRCh38
rs907955076 Chr.7: 65980119-65980119 on Build GRCh38
rs562104880 Chr.7: 65979688-65979688 on Build GRCh38
rs748161388 Chr.7: 65979896-65979896 on Build GRCh38
rs556297556 Chr.7: 65980129-65980129 on Build GRCh38
rs764533508 Chr.7: 65979956-65979956 on Build GRCh38
rs1011541170 Chr.7: 65980076-65980076 on Build GRCh38
rs755465262 Chr.7: 65979755-65979755 on Build GRCh38
rs368816546 Chr.7: 65979677-65979677 on Build GRCh38

View Full Product Details