Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.7: 100627800-100628295 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
TFR2
Entrez Gene ID
7036
Gene Name
transferrin receptor 2
Gene Aliases
HFE3, TFRC2
UniGene
Hs.544932
Gene Chromosome Location
Chr.7: 100620416-100642780 on Build GRCh38
Transcripts
NM_001206855.1 NM_003227.3 XM_005250553.4 XM_017012573.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
6228049 c.1591A>G p.S531G Chr.7: 100627921-100627921 on Build GRCh38
3080553 c.1626T>A p.S542R Chr.7: 100627800-100627800 on Build GRCh38
4645580 c.1625G>A p.S542N Chr.7: 100627801-100627801 on Build GRCh38
4908565 c.1403G>A p.R468H Chr.7: 100628294-100628294 on Build GRCh38
4587003 c.1606G>C p.V536L Chr.7: 100627820-100627820 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs1051822153 Chr.7: 100627916-100627916 on Build GRCh38
rs762935630 Chr.7: 100628229-100628229 on Build GRCh38
rs772489994 Chr.7: 100628006-100628006 on Build GRCh38
rs978913572 Chr.7: 100627888-100627888 on Build GRCh38
rs748149161 Chr.7: 100627863-100627863 on Build GRCh38
rs41295900 Chr.7: 100628094-100628094 on Build GRCh38
rs777948696 Chr.7: 100627941-100627941 on Build GRCh38
rs762832274 Chr.7: 100628092-100628092 on Build GRCh38
rs374113312 Chr.7: 100628210-100628210 on Build GRCh38
rs748905972 Chr.7: 100628264-100628264 on Build GRCh38
rs748026419 Chr.7: 100627983-100627984 on Build GRCh38
rs1040713439 Chr.7: 100628206-100628206 on Build GRCh38
rs772672626 Chr.7: 100628168-100628168 on Build GRCh38
rs1023668837 Chr.7: 100628081-100628081 on Build GRCh38
rs747312334 Chr.7: 100628143-100628143 on Build GRCh38
rs780410371 Chr.7: 100628200-100628200 on Build GRCh38
rs761009757 Chr.7: 100628098-100628098 on Build GRCh38
rs41295899 Chr.7: 100628248-100628248 on Build GRCh38
rs142766112 Chr.7: 100628103-100628103 on Build GRCh38
rs959981080 Chr.7: 100628068-100628068 on Build GRCh38
rs371347559 Chr.7: 100627976-100627976 on Build GRCh38
rs774894320 Chr.7: 100627918-100627918 on Build GRCh38
rs186378421 Chr.7: 100628208-100628208 on Build GRCh38
rs770754691 Chr.7: 100628173-100628173 on Build GRCh38
rs761297359 Chr.7: 100628059-100628059 on Build GRCh38
rs778438972 Chr.7: 100628266-100628266 on Build GRCh38
rs765131429 Chr.7: 100627958-100627958 on Build GRCh38
rs763692373 Chr.7: 100627945-100627945 on Build GRCh38
rs80338886 Chr.7: 100628228-100628228 on Build GRCh38
rs934731307 Chr.7: 100627901-100627901 on Build GRCh38
rs373936655 Chr.7: 100627964-100627964 on Build GRCh38
rs752990792 Chr.7: 100627973-100627973 on Build GRCh38
rs778860645 Chr.7: 100627982-100627983 on Build GRCh38
rs759461055 Chr.7: 100628216-100628216 on Build GRCh38
rs772014889 Chr.7: 100628217-100628217 on Build GRCh38
rs748238420 Chr.7: 100627885-100627885 on Build GRCh38
rs754731616 Chr.7: 100627839-100627840 on Build GRCh38
rs373025420 Chr.7: 100628205-100628205 on Build GRCh38
rs768409892 Chr.7: 100628145-100628145 on Build GRCh38
rs763454180 Chr.7: 100627850-100627850 on Build GRCh38
rs916143659 Chr.7: 100627882-100627882 on Build GRCh38
rs1003928892 Chr.7: 100628001-100628001 on Build GRCh38
rs777401563 Chr.7: 100628259-100628259 on Build GRCh38
rs201746674 Chr.7: 100627858-100627858 on Build GRCh38
rs943359654 Chr.7: 100627968-100627968 on Build GRCh38
rs369755113 Chr.7: 100627920-100627920 on Build GRCh38
rs781475554 Chr.7: 100627884-100627884 on Build GRCh38
rs201943504 Chr.7: 100628083-100628083 on Build GRCh38
rs752025865 Chr.7: 100628203-100628203 on Build GRCh38
rs951124614 Chr.7: 100627846-100627846 on Build GRCh38
rs987977631 Chr.7: 100628115-100628115 on Build GRCh38
rs771822800 Chr.7: 100628213-100628213 on Build GRCh38
rs767245661 Chr.7: 100627827-100627827 on Build GRCh38
rs756218360 Chr.7: 100627895-100627895 on Build GRCh38
rs768160819 Chr.7: 100628036-100628036 on Build GRCh38
rs886905130 Chr.7: 100627993-100627993 on Build GRCh38
rs758179159 Chr.7: 100627971-100627971 on Build GRCh38
rs1049922942 Chr.7: 100628159-100628159 on Build GRCh38
rs777532178 Chr.7: 100628124-100628124 on Build GRCh38
rs202043407 Chr.7: 100628017-100628017 on Build GRCh38
rs533880980 Chr.7: 100628247-100628247 on Build GRCh38
rs144148322 Chr.7: 100628226-100628226 on Build GRCh38
rs377108556 Chr.7: 100627992-100627992 on Build GRCh38
rs200201634 Chr.7: 100628211-100628211 on Build GRCh38
rs201082151 Chr.7: 100628209-100628209 on Build GRCh38
rs148541221 Chr.7: 100628097-100628097 on Build GRCh38
rs771540356 Chr.7: 100627898-100627898 on Build GRCh38
rs377542243 Chr.7: 100628188-100628188 on Build GRCh38
rs376719507 Chr.7: 100627972-100627972 on Build GRCh38
rs759247993 Chr.7: 100628180-100628180 on Build GRCh38
rs747728512 Chr.7: 100628154-100628154 on Build GRCh38
rs890005464 Chr.7: 100627920-100627921 on Build GRCh38
rs754040201 Chr.7: 100628107-100628107 on Build GRCh38
rs374546301 Chr.7: 100628214-100628214 on Build GRCh38
rs760178993 Chr.7: 100627943-100627943 on Build GRCh38
rs758992254 Chr.7: 100628197-100628197 on Build GRCh38
rs779079099 Chr.7: 100628121-100628121 on Build GRCh38
rs373084596 Chr.7: 100627861-100627861 on Build GRCh38
rs377429498 Chr.7: 100627865-100627865 on Build GRCh38
rs369107718 Chr.7: 100627994-100627994 on Build GRCh38
rs931476373 Chr.7: 100628137-100628137 on Build GRCh38
rs767913945 Chr.7: 100628241-100628241 on Build GRCh38
rs766437336 Chr.7: 100628111-100628111 on Build GRCh38
rs373415641 Chr.7: 100628125-100628125 on Build GRCh38
rs1026719197 Chr.7: 100627845-100627845 on Build GRCh38
rs769302506 Chr.7: 100628161-100628161 on Build GRCh38
rs757146818 Chr.7: 100627864-100627864 on Build GRCh38
rs774161209 Chr.7: 100628212-100628212 on Build GRCh38
rs376230804 Chr.7: 100628207-100628207 on Build GRCh38
rs756733318 Chr.7: 100628265-100628265 on Build GRCh38
rs368765129 Chr.7: 100627848-100627848 on Build GRCh38
rs775956952 Chr.7: 100627949-100627949 on Build GRCh38
rs762922296 Chr.7: 100628171-100628171 on Build GRCh38
rs767276889 Chr.7: 100628183-100628183 on Build GRCh38
rs766412097 Chr.7: 100628236-100628236 on Build GRCh38
rs890069610 Chr.7: 100628179-100628179 on Build GRCh38
rs577722163 Chr.7: 100627897-100627897 on Build GRCh38
rs899733813 Chr.7: 100627978-100627978 on Build GRCh38
rs368814331 Chr.7: 100627990-100627990 on Build GRCh38
rs886061833 Chr.7: 100627937-100627937 on Build GRCh38
rs750198791 Chr.7: 100627869-100627869 on Build GRCh38
rs139178017 Chr.7: 100628224-100628224 on Build GRCh38
rs1019233469 Chr.7: 100627944-100627944 on Build GRCh38
rs755236142 Chr.7: 100627877-100627877 on Build GRCh38
rs546954761 Chr.7: 100627875-100627875 on Build GRCh38
rs765439073 Chr.7: 100628191-100628191 on Build GRCh38
rs200053955 Chr.7: 100628082-100628082 on Build GRCh38
rs376917998 Chr.7: 100627826-100627826 on Build GRCh38
rs41295903 Chr.7: 100627986-100627986 on Build GRCh38
rs775385253 Chr.7: 100628222-100628222 on Build GRCh38
rs746704550 Chr.7: 100628025-100628025 on Build GRCh38
rs761790083 Chr.7: 100627957-100627957 on Build GRCh38
rs750609759 Chr.7: 100627822-100627822 on Build GRCh38
rs780814385 Chr.7: 100628148-100628148 on Build GRCh38
rs750499411 Chr.7: 100628192-100628192 on Build GRCh38
rs770286722 Chr.7: 100628195-100628195 on Build GRCh38
rs146530042 Chr.7: 100628237-100628237 on Build GRCh38
rs150618729 Chr.7: 100628106-100628106 on Build GRCh38
rs746317846 Chr.7: 100627997-100627997 on Build GRCh38
rs774262230 Chr.7: 100628174-100628174 on Build GRCh38
rs754890555 Chr.7: 100627835-100627835 on Build GRCh38
rs200503408 Chr.7: 100628039-100628039 on Build GRCh38
rs775222082 Chr.7: 100627923-100627923 on Build GRCh38
rs771908334 Chr.7: 100627921-100627921 on Build GRCh38
rs754126409 Chr.7: 100628185-100628185 on Build GRCh38

View Full Product Details