Assay Design

The assay primers and probes lie within a single exon. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
EBF3
Entrez Gene ID
253738
Gene Name
early B-cell factor 3
Gene Aliases
COE3, EBF-3, O/E-2, OE-2
Chromosome Location
Chr.10: 129835232 - 129964274 on Build GRCh38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001005463.2 NP_001005463.1 16 - 16 3203 78
XM_005252667.3 XP_005252724.1 17 - 17 3871 78
XM_005252668.3 XP_005252725.1 16 - 16 3785 78
XM_005252669.3 XP_005252726.1 16 - 16 3758 78
XM_006717739.3 XP_006717802.1 17 - 17 3898 78
XM_006717740.3 XP_006717803.1 17 - 17 3895 78
XM_006717742.3 XP_006717805.1 17 - 17 3790 78
XM_006717743.3 XP_006717806.1 16 - 16 3677 78
XM_006717744.3 XP_006717807.1 16 - 16 3601 78
XM_011539574.2 XP_011537876.1 15 - 15 3613 78
XM_011539575.2 XP_011537877.1 12 - 12 2892 78
XM_017016027.1 XP_016871516.1 15 - 15 3488 78
GenBank mRNA AK055710.1 - 1 - 1 351 78
AK098025.1 - 1 - 1 964 78

View Full Product Details