Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
SHF
Entrez Gene ID
90525
Gene Name
Src homology 2 domain containing F
Gene Aliases
-
Chromosome Location
Chr.15: 45167214 - 45201175 on Build GRCh38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001301168.1 NP_001288097.1 3 - 4 866 57
NM_001301169.1 NP_001288098.1 3 - 4 866 57
NM_001301171.1 NP_001288100.1 3 - 4 453 57
GenBank mRNA AK301625.1 - 3 - 4 453 57
AK303749.1 - 4 - 5 641 57
BC007586.1 - 2 - 3 228 57
BC171730.1 - 2 - 3 306 57

View Full Product Details