Assay Design
The assay primers and probes lie within a single exon. This assay will detect gDNA.
Gene Transcripts
- Gene Symbol
- FBRSL1
- Entrez Gene ID
-
57666
- Gene Name
- fibrosin like 1
- Gene Aliases
-
-
- Chromosome Location
-
Chr.12: 132490440 - 132585188 on Build GRCh38
Non Gene Transcripts
Interrogated Sequence |
Translated Protein |
Exon Boundary |
Assay Location |
IMAGE Clone ID |
Transcript Location |
Amplicon Length |
GenBankmRNA |
BE737149.1 |
- |
1 |
372 |
|
Chr. 12 - 132513798 - 132514595 [+] on Build GRCh38 |
77 |
GenBankmRNA |
CN367605.1 |
- |
1 |
357 |
|
Chr. 12 - 132513790 - 132514314 [+] on Build GRCh38 |
77 |
GenBankmRNA |
BM724031.1 |
- |
1 |
285 |
|
Chr. 12 - 132513847 - 132514494 [+] on Build GRCh38 |
77 |
Adjacent Gene Details
|
Gene Name |
Gene Symbol |
Alias |
Distance |
Relative Direction |
Location Chromosome |
Upstream |
mucin 8 |
MUC8 |
|
-39.2kb
|
Anti-sense |
Chr. - 132471092 - 132476638 on Build GRCh38 |
Downstream |
WAS/WASL-interacting protein family member 3-like |
LOC100996573 |
|
35.2kb
|
Sense |
Chr. - 132550729 - 132559465 on Build GRCh38 |
View Full Product Details