The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_001099777.3 | NP_001093247.1 | 2 - 3 | 277 | 76 |
NM_001130001.2 | NP_001123473.1 | 2 - 3 | 277 | 76 | |
NM_001130002.2 | NP_001123474.1 | 2 - 3 | 277 | 76 | |
NM_001243729.1 | NP_001230658.1 | 2 - 3 | 277 | 76 | |
XM_017005754.1 | XP_016861243.1 | 2 - 3 | 231 | 76 | |
GenBank mRNA | BC010062.2 | - | 2 - 3 | 177 | 76 |
BC029129.1 | - | 2 - 3 | 224 | 76 | |
BC126187.1 | - | 2 - 3 | 244 | 76 | |
BC126189.1 | - | 2 - 3 | 244 | 76 | |
BC144066.1 | - | 2 - 3 | 244 | 76 | |
BG217991.1 | - | 1 - 2 | 111 | 76 |