The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_138424.1 | NP_612433.1 | 4 - 5 | 473 | 61 |
XM_005251683.4 | XP_005251740.1 | 7 - 8 | 1462 | 61 | |
XM_006716947.2 | XP_006717010.1 | 6 - 7 | 714 | 61 | |
XM_011518190.2 | XP_011516492.1 | 7 - 8 | 1497 | 61 | |
XM_011518191.2 | XP_011516493.1 | 7 - 8 | 1496 | 61 | |
XM_011518192.2 | XP_011516494.1 | 7 - 8 | 1494 | 61 | |
XM_011518193.2 | XP_011516495.1 | 7 - 8 | 1504 | 61 | |
GenBank mRNA | BC010626.1 | - | 4 - 5 | 473 | 61 |