The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_173546.2 | NP_775817.1 | 5 - 6 | 1108 | 97 |
XM_005264938.2 | XP_005264995.1 | 5 - 6 | 1594 | 97 | |
XM_005264940.4 | XP_005264997.1 | 5 - 6 | 1229 | 97 | |
XM_006713015.2 | XP_006713078.1 | 5 - 6 | 1624 | 97 | |
XM_006713016.2 | XP_006713079.1 | 5 - 6 | 1603 | 97 | |
GenBank mRNA | AK294160.1 | - | 5 - 6 | 957 | 97 |
AY129011.1 | - | 5 - 6 | 1065 | 97 | |
BC013110.2 | - | 1 - 2 | 102 | 97 | |
BC039083.1 | - | 5 - 6 | 1058 | 97 |