Assay Design

The assay primers and probes lie within a single exon. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
NCAM2
Entrez Gene ID
4685
Gene Name
neural cell adhesion molecule 2
Gene Aliases
NCAM21
Chromosome Location
Chr.21: 20998315 - 21543324 on Build GRCh38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq XM_011529575.2 XP_011527877.1 19 - 19 5728 64
XM_011529576.2 XP_011527878.1 19 - 19 5305 64
XM_011529579.2 XP_011527881.1 18 - 18 5229 64
XM_011529585.2 XP_011527887.1 15 - 15 4595 64
XM_017028353.1 XP_016883842.1 19 - 19 5357 64
XM_017028356.1 XP_016883845.1 18 - 18 5128 64
XM_017028357.1 XP_016883846.1 18 - 18 5092 64
GenBank mRNA AL137344.1 - 1 - 1 253 64

View Full Product Details