Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.X: 18650303-18650795 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
CDKL5
Entrez Gene ID
6792
Gene Name
cyclin dependent kinase like 5
Gene Aliases
CFAP247, EIEE2, ISSX, STK9
UniGene
Hs.659851
Gene Chromosome Location
Chr.X: 18425605-18653629 on Build GRCh38
Transcripts
NM_001037343.1 NM_003159.2 NM_000330.3

Gene Symbol
RS1
Entrez Gene ID
6247
Gene Name
retinoschisin 1
Gene Aliases
RS, XLRS1
UniGene
Hs.715725
Gene Chromosome Location
Chr.X: 18639688-18672103 on Build GRCh38
Transcripts
NM_001037343.1 NM_003159.2 NM_000330.3

SNP Detail

SNP ID Chromosome Location
rs750449476 Chr.X: 18650487-18650487 on Build GRCh38
rs752120798 Chr.X: 18650491-18650491 on Build GRCh38
rs774335258 Chr.X: 18650457-18650457 on Build GRCh38
rs769305518 Chr.X: 18650511-18650511 on Build GRCh38
rs368135097 Chr.X: 18650639-18650639 on Build GRCh38
rs371847235 Chr.X: 18650488-18650488 on Build GRCh38
rs368344738 Chr.X: 18650453-18650453 on Build GRCh38
rs570887192 Chr.X: 18650521-18650521 on Build GRCh38
rs140944590 Chr.X: 18650540-18650540 on Build GRCh38
rs587783403 Chr.X: 18650432-18650432 on Build GRCh38
rs367896650 Chr.X: 18650356-18650356 on Build GRCh38
rs371225621 Chr.X: 18650367-18650367 on Build GRCh38
rs998955164 Chr.X: 18650743-18650743 on Build GRCh38
rs953726340 Chr.X: 18650476-18650476 on Build GRCh38
rs372022123 Chr.X: 18650640-18650640 on Build GRCh38
rs999132676 Chr.X: 18650362-18650362 on Build GRCh38
rs781435432 Chr.X: 18650502-18650502 on Build GRCh38
rs980932278 Chr.X: 18650390-18650390 on Build GRCh38
rs142949334 Chr.X: 18650771-18650771 on Build GRCh38
rs998058972 Chr.X: 18650391-18650391 on Build GRCh38
rs374054249 Chr.X: 18650553-18650553 on Build GRCh38
rs767312604 Chr.X: 18650469-18650469 on Build GRCh38
rs376557374 Chr.X: 18650479-18650479 on Build GRCh38
rs777709363 Chr.X: 18650353-18650353 on Build GRCh38
rs765535216 Chr.X: 18650483-18650483 on Build GRCh38
rs866859766 Chr.X: 18650592-18650592 on Build GRCh38
rs190555376 Chr.X: 18650368-18650368 on Build GRCh38
rs780702363 Chr.X: 18650489-18650489 on Build GRCh38
rs267608665 Chr.X: 18650520-18650520 on Build GRCh38
rs775114662 Chr.X: 18650547-18650547 on Build GRCh38
rs200236257 Chr.X: 18650442-18650442 on Build GRCh38
rs376982458 Chr.X: 18650359-18650359 on Build GRCh38
rs587783161 Chr.X: 18650539-18650539 on Build GRCh38
rs892437453 Chr.X: 18650668-18650669 on Build GRCh38
rs1044040565 Chr.X: 18650740-18650740 on Build GRCh38
rs772632048 Chr.X: 18650392-18650392 on Build GRCh38
rs148531754 Chr.X: 18650613-18650613 on Build GRCh38
rs1040778236 Chr.X: 18650658-18650658 on Build GRCh38
rs764791214 Chr.X: 18650510-18650542 on Build GRCh38
rs760568446 Chr.X: 18650554-18650554 on Build GRCh38
rs904637400 Chr.X: 18650672-18650672 on Build GRCh38
rs772246921 Chr.X: 18650611-18650611 on Build GRCh38
rs904087104 Chr.X: 18650750-18650750 on Build GRCh38
rs770428990 Chr.X: 18650531-18650531 on Build GRCh38
rs202153551 Chr.X: 18650466-18650466 on Build GRCh38
rs1009561916 Chr.X: 18650736-18650736 on Build GRCh38
rs143243059 Chr.X: 18650545-18650545 on Build GRCh38
rs752986338 Chr.X: 18650633-18650633 on Build GRCh38
rs765366935 Chr.X: 18650570-18650570 on Build GRCh38
rs745659914 Chr.X: 18650609-18650609 on Build GRCh38
rs1057408898 Chr.X: 18650599-18650599 on Build GRCh38
rs865984450 Chr.X: 18650538-18650538 on Build GRCh38
rs181859624 Chr.X: 18650612-18650612 on Build GRCh38
rs747799506 Chr.X: 18650508-18650508 on Build GRCh38
rs750725714 Chr.X: 18650591-18650591 on Build GRCh38

View Full Product Details