Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.X: 41474929-41475442 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
NYX
Entrez Gene ID
60506
Gene Name
nyctalopin
Gene Aliases
CLRP, CSNB1, CSNB1A, CSNB4, NBM1
UniGene
Hs.302019
Gene Chromosome Location
Chr.X: 41447460-41476414 on Build GRCh38
Transcripts
NM_022567.2 XM_017029709.1

SNP Detail

SNP ID Chromosome Location
rs767300031 Chr.X: 41475287-41475287 on Build GRCh38
rs978067833 Chr.X: 41475026-41475026 on Build GRCh38
rs921009553 Chr.X: 41475303-41475303 on Build GRCh38
rs889858342 Chr.X: 41475100-41475100 on Build GRCh38
rs995740946 Chr.X: 41475251-41475251 on Build GRCh38
rs3021319 Chr.X: 41475408-41475408 on Build GRCh38
rs148699627 Chr.X: 41475282-41475282 on Build GRCh38
rs765627273 Chr.X: 41475099-41475099 on Build GRCh38
rs891839220 Chr.X: 41475029-41475029 on Build GRCh38
rs1019733370 Chr.X: 41475335-41475335 on Build GRCh38
rs367599214 Chr.X: 41475143-41475143 on Build GRCh38
rs762976224 Chr.X: 41475194-41475194 on Build GRCh38
rs753101555 Chr.X: 41475047-41475048 on Build GRCh38
rs976932884 Chr.X: 41475285-41475285 on Build GRCh38
rs978120347 Chr.X: 41475354-41475354 on Build GRCh38
rs761926242 Chr.X: 41474960-41474960 on Build GRCh38
rs1057515883 Chr.X: 41474971-41474971 on Build GRCh38
rs750048192 Chr.X: 41475002-41475002 on Build GRCh38
rs747097738 Chr.X: 41475416-41475416 on Build GRCh38
rs952518001 Chr.X: 41475197-41475197 on Build GRCh38
rs935165720 Chr.X: 41475306-41475306 on Build GRCh38
rs1007359869 Chr.X: 41475109-41475109 on Build GRCh38
rs906348045 Chr.X: 41474993-41474993 on Build GRCh38
rs370733549 Chr.X: 41475051-41475051 on Build GRCh38
rs965450101 Chr.X: 41475338-41475338 on Build GRCh38
rs916377636 Chr.X: 41475117-41475117 on Build GRCh38
rs187741852 Chr.X: 41475006-41475006 on Build GRCh38
rs1052338157 Chr.X: 41475365-41475365 on Build GRCh38
rs947814333 Chr.X: 41475133-41475133 on Build GRCh38
rs972982122 Chr.X: 41475188-41475188 on Build GRCh38
rs923893223 Chr.X: 41475044-41475044 on Build GRCh38
rs58930096 Chr.X: 41475313-41475313 on Build GRCh38
rs550775335 Chr.X: 41475198-41475198 on Build GRCh38
rs1025898413 Chr.X: 41475191-41475191 on Build GRCh38
rs751371521 Chr.X: 41475203-41475203 on Build GRCh38
rs755307939 Chr.X: 41475012-41475012 on Build GRCh38
rs893711795 Chr.X: 41475374-41475374 on Build GRCh38
rs927806332 Chr.X: 41475297-41475297 on Build GRCh38
rs767111588 Chr.X: 41474962-41474962 on Build GRCh38
rs906280880 Chr.X: 41475158-41475158 on Build GRCh38
rs753104188 Chr.X: 41475171-41475171 on Build GRCh38
rs531963566 Chr.X: 41475014-41475014 on Build GRCh38
rs1011570912 Chr.X: 41475039-41475039 on Build GRCh38
rs371864261 Chr.X: 41475218-41475218 on Build GRCh38
rs1057515884 Chr.X: 41475056-41475056 on Build GRCh38

View Full Product Details