Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.1: 202940566-202941102 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
ADIPOR1
Entrez Gene ID
51094
Gene Name
adiponectin receptor 1
Gene Aliases
ACDCR1, CGI-45, CGI45, PAQR1, TESBP1A
UniGene
Hs.5298
Gene Chromosome Location
Chr.1: 202940825-202958572 on Build GRCh38
Transcripts
NM_001290553.1 NM_001290557.1 NM_001290629.1 NM_015999.5 XM_017001407.1

SNP Detail

SNP ID Chromosome Location
rs960388097 Chr.1: 202940595-202940595 on Build GRCh38
rs750572117 Chr.1: 202940911-202940911 on Build GRCh38
rs757482146 Chr.1: 202940760-202940760 on Build GRCh38
rs544030104 Chr.1: 202940800-202940800 on Build GRCh38
rs974571873 Chr.1: 202940897-202940897 on Build GRCh38
rs747941880 Chr.1: 202940949-202940949 on Build GRCh38
rs1033925056 Chr.1: 202940810-202940810 on Build GRCh38
rs950414036 Chr.1: 202940864-202940864 on Build GRCh38
rs984580920 Chr.1: 202940886-202940886 on Build GRCh38
rs929118164 Chr.1: 202940682-202940682 on Build GRCh38
rs551634906 Chr.1: 202940948-202940948 on Build GRCh38
rs934020471 Chr.1: 202940692-202940692 on Build GRCh38
rs943472462 Chr.1: 202940607-202940607 on Build GRCh38
rs923060558 Chr.1: 202940924-202940924 on Build GRCh38
rs534570951 Chr.1: 202940606-202940606 on Build GRCh38
rs1005923023 Chr.1: 202940725-202940725 on Build GRCh38
rs8167 Chr.1: 202941059-202941059 on Build GRCh38
rs887692337 Chr.1: 202940719-202940719 on Build GRCh38
rs892427121 Chr.1: 202940701-202940701 on Build GRCh38
rs1044091839 Chr.1: 202940736-202940736 on Build GRCh38
rs80165154 Chr.1: 202940993-202940993 on Build GRCh38
rs773862347 Chr.1: 202941031-202941031 on Build GRCh38
rs11547945 Chr.1: 202940981-202940981 on Build GRCh38
rs991775678 Chr.1: 202940595-202940595 on Build GRCh38
rs1016414254 Chr.1: 202940891-202940891 on Build GRCh38
rs913920560 Chr.1: 202940981-202940982 on Build GRCh38
rs879234012 Chr.1: 202940982-202940982 on Build GRCh38
rs1051098549 Chr.1: 202940693-202940693 on Build GRCh38
rs879248167 Chr.1: 202940774-202940774 on Build GRCh38
rs533494322 Chr.1: 202941037-202941037 on Build GRCh38
rs574544277 Chr.1: 202940610-202940610 on Build GRCh38
rs113890019 Chr.1: 202941050-202941050 on Build GRCh38
rs540758512 Chr.1: 202940859-202940859 on Build GRCh38
rs968043667 Chr.1: 202940603-202940603 on Build GRCh38
rs56934600 Chr.1: 202940609-202940609 on Build GRCh38
rs1025952022 Chr.1: 202940856-202940856 on Build GRCh38
rs988698175 Chr.1: 202940980-202940980 on Build GRCh38
rs1047664913 Chr.1: 202940686-202940686 on Build GRCh38
rs112936913 Chr.1: 202940932-202940932 on Build GRCh38
rs7539542 Chr.1: 202940846-202940846 on Build GRCh38
rs964610310 Chr.1: 202940896-202940896 on Build GRCh38
rs904042515 Chr.1: 202940754-202940754 on Build GRCh38
rs543050405 Chr.1: 202940813-202940813 on Build GRCh38
rs756434162 Chr.1: 202940887-202940887 on Build GRCh38
rs1043843230 Chr.1: 202940990-202940990 on Build GRCh38
rs572098528 Chr.1: 202940605-202940605 on Build GRCh38
rs189663347 Chr.1: 202940881-202940881 on Build GRCh38

View Full Product Details