Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.11: 47216244-47216737 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
DDB2
Entrez Gene ID
1643
Gene Name
damage specific DNA binding protein 2
Gene Aliases
DDBB, UV-DDB2, XPE
UniGene
Hs.700338
Gene Chromosome Location
Chr.11: 47214942-47239218 on Build GRCh38
Transcripts
NM_000107.2 NM_001300734.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
1249843 c.210C>T p.V70V Chr.11: 47216418-47216418 on Build GRCh38
1298086 c.158T>C p.L53P Chr.11: 47216366-47216366 on Build GRCh38
6385218 c.172G>A p.A58T Chr.11: 47216380-47216380 on Build GRCh38
3809303 c.189G>A p.L63L Chr.11: 47216397-47216397 on Build GRCh38
5903778 c.227A>C p.H76P Chr.11: 47216435-47216435 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs749426163 Chr.11: 47216445-47216445 on Build GRCh38
rs772461740 Chr.11: 47216489-47216489 on Build GRCh38
rs910867251 Chr.11: 47216536-47216536 on Build GRCh38
rs535591210 Chr.11: 47216350-47216350 on Build GRCh38
rs748397343 Chr.11: 47216327-47216327 on Build GRCh38
rs904410426 Chr.11: 47216466-47216466 on Build GRCh38
rs770922074 Chr.11: 47216462-47216462 on Build GRCh38
rs758139129 Chr.11: 47216320-47216320 on Build GRCh38
rs1014697958 Chr.11: 47216621-47216621 on Build GRCh38
rs777824005 Chr.11: 47216432-47216432 on Build GRCh38
rs759879517 Chr.11: 47216355-47216355 on Build GRCh38
rs191773047 Chr.11: 47216397-47216397 on Build GRCh38
rs777879172 Chr.11: 47216333-47216333 on Build GRCh38
rs568187738 Chr.11: 47216405-47216405 on Build GRCh38
rs549788260 Chr.11: 47216325-47216325 on Build GRCh38
rs150620642 Chr.11: 47216374-47216374 on Build GRCh38
rs181404957 Chr.11: 47216580-47216580 on Build GRCh38
rs374456936 Chr.11: 47216290-47216290 on Build GRCh38
rs758168272 Chr.11: 47216415-47216415 on Build GRCh38
rs553748511 Chr.11: 47216514-47216514 on Build GRCh38
rs952673674 Chr.11: 47216714-47216714 on Build GRCh38
rs993991303 Chr.11: 47216658-47216658 on Build GRCh38
rs745735733 Chr.11: 47216302-47216302 on Build GRCh38
rs774817566 Chr.11: 47216268-47216268 on Build GRCh38
rs3740682 Chr.11: 47216698-47216698 on Build GRCh38
rs1008710405 Chr.11: 47216535-47216535 on Build GRCh38
rs890276220 Chr.11: 47216532-47216532 on Build GRCh38
rs994876446 Chr.11: 47216581-47216581 on Build GRCh38
rs983715198 Chr.11: 47216604-47216604 on Build GRCh38
rs199630230 Chr.11: 47216426-47216426 on Build GRCh38
rs772355051 Chr.11: 47216366-47216366 on Build GRCh38
rs763720169 Chr.11: 47216286-47216286 on Build GRCh38
rs916643112 Chr.11: 47216700-47216700 on Build GRCh38
rs755646110 Chr.11: 47216276-47216276 on Build GRCh38
rs1035300098 Chr.11: 47216657-47216657 on Build GRCh38
rs749763301 Chr.11: 47216337-47216337 on Build GRCh38
rs778571431 Chr.11: 47216295-47216295 on Build GRCh38
rs576196809 Chr.11: 47216628-47216628 on Build GRCh38
rs4647710 Chr.11: 47216595-47216596 on Build GRCh38
rs895786891 Chr.11: 47216609-47216609 on Build GRCh38
rs943716869 Chr.11: 47216538-47216538 on Build GRCh38
rs1026898559 Chr.11: 47216529-47216529 on Build GRCh38
rs756368265 Chr.11: 47216431-47216431 on Build GRCh38
rs779731799 Chr.11: 47216416-47216416 on Build GRCh38
rs776041354 Chr.11: 47216504-47216504 on Build GRCh38
rs369552849 Chr.11: 47216328-47216328 on Build GRCh38
rs897948691 Chr.11: 47216655-47216655 on Build GRCh38
rs950919063 Chr.11: 47216594-47216594 on Build GRCh38
rs1015435321 Chr.11: 47216413-47216413 on Build GRCh38
rs946249161 Chr.11: 47216278-47216278 on Build GRCh38
rs976516916 Chr.11: 47216540-47216540 on Build GRCh38
rs753467757 Chr.11: 47216400-47216400 on Build GRCh38
rs140354834 Chr.11: 47216693-47216693 on Build GRCh38
rs757304535 Chr.11: 47216291-47216291 on Build GRCh38
rs1017771819 Chr.11: 47216573-47216573 on Build GRCh38
rs1043766285 Chr.11: 47216457-47216457 on Build GRCh38
rs144744092 Chr.11: 47216593-47216594 on Build GRCh38
rs760456614 Chr.11: 47216391-47216391 on Build GRCh38
rs186870004 Chr.11: 47216270-47216270 on Build GRCh38
rs771533958 Chr.11: 47216346-47216346 on Build GRCh38
rs185687113 Chr.11: 47216582-47216582 on Build GRCh38
rs374094218 Chr.11: 47216480-47216480 on Build GRCh38
rs141990819 Chr.11: 47216338-47216338 on Build GRCh38
rs139674102 Chr.11: 47216408-47216408 on Build GRCh38
rs752297564 Chr.11: 47216271-47216271 on Build GRCh38
rs960846595 Chr.11: 47216694-47216694 on Build GRCh38
rs913007826 Chr.11: 47216552-47216552 on Build GRCh38
rs765161300 Chr.11: 47216407-47216407 on Build GRCh38
rs983577243 Chr.11: 47216303-47216303 on Build GRCh38
rs577230400 Chr.11: 47216531-47216531 on Build GRCh38

View Full Product Details